Merck
  • Gram scale synthesis of 3-fluoro-1-hydroxyacetone phosphate: a novel donor substrate in rabbit muscle aldolase-catalyzed aldol reactions.
Gram scale synthesis of 3-fluoro-1-hydroxyacetone phosphate: a novel donor substrate in rabbit muscle aldolase-catalyzed aldol reactions.
Chemical communications (Cambridge, England) (2011-05-13)
Michael Fischer, Hanspeter Kählig, Walther Schmid
ABSTRACT
An efficient gram scale synthesis of 3-fluoro-1-hydroxyacetone phosphate (FHAP) has been developed. As a close analog to dihydroxyacetone phosphate, FHAP was used as a novel donor substrate for rabbit muscle aldolase catalyzed reactions. The different binding affinities of the gem-diol and keto form of FHAP were studied by (19)F-NMR.
Sigma-Aldrich
Dihydroxyacetone phosphate hemimagnesium salt hydrate, ≥95% (TLC)