Merck
  • Assessment of fumonisins B1 and B2 levels in commercial maize-based food products by liquid chromatography with fluorimetric detection and postcolumn chemical derivatization.
Assessment of fumonisins B1 and B2 levels in commercial maize-based food products by liquid chromatography with fluorimetric detection and postcolumn chemical derivatization.
Journal of food science (2011-05-04)
Sonia Lo Magro, Maria Campaniello, Donatella Nardiello, Marilena Muscarella
ABSTRACT
The occurrence of the fumonisins B(1) and B(2) in maize-based food products marketed in Italy was examined. A simply and reliable chromatographic method with fluorimetric detection and postcolumn o-phtalaldehyde derivatization was used for a monitoring of 100 samples (8 flours, 21 corn-meal, 16 snacks, 7 maize samples, 13 gluten-free products, and 35 corn-flakes) bought in local supermarkets during the years 2008 and 2009. The presence of both fumonisins B(1) and B(2), at a concentration higher than 15 μg/kg, was observed in all samples of corn-meal and maize-flour, in 75% of snacks, in 57% of maize samples, in 54% of gluten-free products, and in 29% of corn-flakes. A total of 7 samples including 4 corn-meals, 2 maize-flours, and 1 maize showed a value exceeding the maximum level fixed in the Regulation 1126/2007/EC; no positive sample was observed in corn-flakes, snacks, and gluten-free foods. Fumonisins contamination, on the whole range of maize-based food products analyzed, emphasizes the need of improve agricultural practices, and increase official control and monitoring studies.
Sigma-Aldrich
Fumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor