Merck
  • [Biosynthesis of 4,15-diacetylnivalenol by Fusarium sambucinum Fuckel var. minus].
[Biosynthesis of 4,15-diacetylnivalenol by Fusarium sambucinum Fuckel var. minus].
Prikladnaia biokhimiia i mikrobiologiia (2011-03-30)
G D Sokolova, V N Voznesenskiĭ
ABSTRACT
Fusarium sambucinum Fuckel var. minus isolate produced unusual for F. sambucinum Fuckel trichothecene metabolite 4,15-diacetylnivalenol (9 mg/l) in conditions of deep cultivation on Myro medium. This compound was identified by TLC, GLC, HPLC, and 1N MR spectroscopy. Other trichothecenes, 4-acetylnivalenol (3 mg/l) and nivalenol (1 mg/l), were also found in the culture. The observed feature of the studied isolate is assumed to be due to the presence of an additional gene, which encodes cytochrome P450 oxygenase responsible for the introduction of keto group at C8 and hydroxyl group at C7 of the trichothecene structure.
Product Description
Supelco
Nivalenol solution, 100 μg/mL in acetonitrile, analytical standard
Nivalenol in acetonitrile, IRMM®, certified reference material