Merck
Ytterbium trifluoride as a radiopaque agent for dental cements.
International endodontic journal (2010-06-29)
F M Collares, F A Ogliari, G S Lima, V R C Fontanella, E Piva, S M W Samuel
ABSTRACT
To evaluate the radiopacity, degree of conversion (DC) and flexural strength of an experimental dental cement, with several added radiopaque substances. Titanium dioxide, quartz, zirconia, bismuth oxide, barium sulphate and ytterbium trifluoride were added to the experimental cement in five different concentrations. Radiopacity was evaluated with a phosphor plate system, and the radiodensity of specimens was compared with an aluminium step-wedge. DC was evaluated with FT-infrared spectroscopy following 20 s of photo-activation. Specimens with dimensions of 12 x 2 x 2 mm were used for the flexural strength test. Data were analysed with two-way anova and Tukey's post hoc test. Radiopacity of the experimental dental cements with barium sulphate and bismuth oxide at 40% and ytterbium fluoride at 30% and 40% showed no significant differences in comparison with 3 mm of Al (181, 96). The experimental dental cements with at least 30% added ytterbium trifluoride had satisfactory radiopacity without influencing other properties.
Sigma-Aldrich
Ytterbium(III) fluoride, anhydrous, powder, 99.98% trace metals basis
Sigma-Aldrich
(1S)-(+)-Camphorquinone, 99%
Sigma-Aldrich
Ethyl 4-(dimethylamino)benzoate, ≥99%
Supelco
Ethyl 4-(dimethylamino)benzoate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland