Merck
  • [Dose-effect relationship of para-toluenesulfonamide for treatment of hepatocellular carcinoma in rats].
[Dose-effect relationship of para-toluenesulfonamide for treatment of hepatocellular carcinoma in rats].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2008-02-06)
Min-ying Li, Hui Meng, Wei-liang Zhu, Shao-zhang Zhou, Hong-yi Li, Ji-ren Zhang
ABSTRACT
To investigate the dose-effect relationship of para-toluenesulfonamide (PTS) for treatment of hepatocellular carcinoma in rats. Forty-two SD rats bearing subcutaneous transplanted hepatocellular carcinoma were randomly divided into 6 groups (n=7), in which 0.02, 0.04, 0.06, 0.08, and 0.10 ml PTS and 0.10 ml normal saline were injected into the tumor, respectively. All of the rats were executed 24 h after the injection to observe the pathological changes in the tumor. In rats with saline injection, the tumor tissues exhibited no obvious changes and the tumor cells retained the active proliferation. PTS, in contrast, caused coagulation necrosis of the tumor tissue, and the necrotic area expanded with the increase of the injected doses. The necrotic volume of the tumor was in roughly linear correlation with the dose of PTS injected, with the linear regression equation of V (cm(3))=-0.018+2.595Y (where V represents tumor necrosis volume, and Y the injected dose of PTS). The dose-effect relationship of PTS is roughly linear, and the PTS dose for injection can be estimated according to the diameter of the tumor.
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%