Merck
  • Auxin and ethylene interactions control mitotic activity of the quiescent centre, root cap size, and pattern of cap cell differentiation in maize.
Auxin and ethylene interactions control mitotic activity of the quiescent centre, root cap size, and pattern of cap cell differentiation in maize.
Plant, cell & environment (2005-07-14)
Georgina Ponce, Peter W Barlow, Lewis J Feldman, Gladys I Cassab
ABSTRACT
Root caps (RCs) are the terminal tissues of higher plant roots. In the present study the factors controlling RC size, shape and structure were examined. It was found that this control involves interactions between the RC and an adjacent population of slowly dividing cells, the quiescent centre, QC. Using the polar auxin transport inhibitor 1-N-naphthylphthalamic acid (NPA), the effects of QC activation on RC gene expression and border cell release was characterized. Ethylene was found to regulate RC size and cell differentiation, since its addition, or the inhibition of its synthesis, affected RC development. The stimulation of cell division in the QC following NPA treatment was reversed by ethylene, and quiescence was re-established. Moreover, inhibition of both ethylene synthesis and auxin polar transport triggered a new pattern of cell division in the root epidermis and led to the appearance of supernumerary epidermal cell files with cap-like characteristics. The data suggest that the QC ensures an ordered internal distribution of auxin, and thereby regulates not only the planes of growth and division in both the root apex proper and the RC meristem, but also regulates cell fate in the RC. Ethylene appears to regulate the auxin redistribution system that resides in the RC. Experiments with Arabidopsis roots also reveal that ethylene plays an important role in regulating the auxin sink, and consequently cell fate in the RC.
Sigma-Aldrich
DL-5-Hydroxylysine hydrochloride