Merck
  • Conversion of pyrrole to pyrrole-2-carboxylate by cells of Bacillus megaterium in supercritical CO2.
Conversion of pyrrole to pyrrole-2-carboxylate by cells of Bacillus megaterium in supercritical CO2.
Chemical communications (Cambridge, England) (2002-09-21)
T Matsuda, Y Ohashi, T Harada, R Yanagihara, T Nagasawa, K Nakamura
ABSTRACT
Pyrrole was converted to pyrrole-2-carboxylate in supercritical CO2 using cells of Bacillus megaterium PYR 2910, and the yield of the carboxylation reaction in supercritical CO2 was 12 times higher than that under atmospheric pressure.
Sigma-Aldrich
Pyrrole-2-carboxylic acid, 99%