Merck
  • Segmental allergen challenge enhances chitinase activity and levels of CCL18 in mild atopic asthma.
Segmental allergen challenge enhances chitinase activity and levels of CCL18 in mild atopic asthma.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2013-01-22)
M L Gavala, E A B Kelly, S Esnault, S Kukreja, M D Evans, P J Bertics, G L Chupp, N N Jarjour
ABSTRACT
Allergic airway inflammation contributes to the airway remodelling that has been linked to increased obstruction and morbidity in asthma. However, the mechanisms by which allergens contribute to airway remodelling in humans are not fully established. CCL18, chitotriosidase (CHIT1) and YKL-40 are readily detectable in the lungs and contribute to remodelling in other fibrotic diseases, but their involvement in allergic asthma is unclear. We hypothesized that CCL18, YKL-40 and CHIT1 bioactivity are enhanced in allergic asthma subjects after segmental allergen challenge and are related to increased pro-fibrotic and Th2-associated mediators in the lungs. Levels of CCL18 and YKL-40 protein and chitotriosidase (CHIT1) bioactivity in bronchoalveolar lavage (BAL) fluid, as well as CCL18, YKL-40 and CHIT1 mRNA levels in BAL cells were evaluated in patients with asthma at baseline and 48 h after segmental allergen challenge. We also examined the correlation between CCL18 and YKL-40 levels and CHIT1 activity with the levels of other pro-fibrotic factors and chemokines previously shown to be up-regulated after allergen challenge. Chitotriosidase activity and YKL-40 and CCL18 levels were elevated after segmental allergen challenge and these levels correlated with those of other pro-fibrotic factors, T cell chemokines, and inflammatory cells after allergen challenge. CCL18 and YKL-40 mRNA levels also increased in BAL cells after allergen challenge. Our results suggest that CCL18 and YKL-40 levels and CHIT1 activity are enhanced in allergic airway inflammation and thus may contribute to airway remodelling in asthma.
Sigma-Aldrich
Chitinase from Streptomyces griseus, lyophilized powder (essentially salt free), ≥200 units/g solid
Sigma-Aldrich
Chitinase from Trichoderma viride, lyophilized powder, ≥600 units/g solid