Merck
  • A sensitive cell-based assay for testing potency of Botulinum neurotoxin type A.
A sensitive cell-based assay for testing potency of Botulinum neurotoxin type A.
ALTEX (2024-07-29)
Ceyda Caliskan, Deniz Simsek, Charlotte Leese, Ciara Doran, Elizabeth Seward, Andrew A Peden, Bazbek Davletov
ABSTRACT
Botulinum neurotoxin type A (BoNT/A) is a widely used biopharmaceutic for the treatment of neurological diseases and aesthetic medicine, allowing months-long paralysis of target muscles and glands. Large numbers of mice are used for multiple botulinum applications including batch release potency testing, antitoxin testing, countermeasure development and basic research. The mouse bioassay (MBA) has historically been the industry gold-standard in the botulinum field and is still heavily used for commercial product testing. BoNT/A intoxication causes severe suffering and application-specific, non-animal alternatives are urgently needed. It is widely accepted, that a cell-based assay (CBA) is the only way to faithfully replicate all the physiological steps of botulinum intoxication; comprising neuronal binding, internalization, endosomal escape, and cleavage of synaptosomal-associated protein of 25 kDa (SNAP25). However, it has not been straightforward to develop these assays and there are only a limited number of CBA currently in use. This is in part, due to the fact that very few cell lines have the appropriate levels of sensitivity to BoNT/A. In this study we have identified that LAN5 cells, a human neuroblastoma derived cell line, are sensitive to BoNT/A and can be engineered to express a recombinant NanoLuc luciferase tagged SNAP25 reporter molecule. On intoxication, the reporter molecule is cleaved and releases a NanoLuc-SNAP25 fragment which can be specifically captured on a 96-well plate for quantitative luminometry. Importantly, we demonstrate this new cell-based assay exhibits sensitivity comparable to the MBA.
Sigma-Aldrich
Anti-GT1b Ganglioside Antibody, ascites fluid, clone GT1b-2b, Chemicon®