Merck
The Role of Uric Acid in Human Health: Insights from the Uricase Gene.
Journal of personalized medicine (2023-09-28)
Youssef M Roman
ABSTRACT
Uric acid is the final product of purine metabolism and is converted to allantoin in most mammals via the uricase enzyme. The accumulation of loss of function mutations in the uricase gene rendered hominoids (apes and humans) to have higher urate concentrations compared to other mammals. The loss of human uricase activity may have allowed humans to survive environmental stressors, evolution bottlenecks, and life-threatening pathogens. While high urate levels may contribute to developing gout and cardiometabolic disorders such as hypertension and insulin resistance, low urate levels may increase the risk for neurodegenerative diseases. The double-edged sword effect of uric acid has resurrected a growing interest in urate's antioxidant role and the uricase enzyme's role in modulating the risk of obesity. Characterizing both the effect of uric acid levels and the uricase enzyme in different animal models may provide new insights into the potential therapeutic benefits of uric acid and novel uricase-based therapy.
Sigma-Aldrich
Uricase from Bacillus fastidiosus, lyophilized, ~9 U/mg