Merck
  • Iron Bioavailability in the Extracellular Environment Is More Relevant Than the Intracellular One in Viability and Gene Expression: A Lesson from Oligodendroglioma Cells.
Iron Bioavailability in the Extracellular Environment Is More Relevant Than the Intracellular One in Viability and Gene Expression: A Lesson from Oligodendroglioma Cells.
Biomedicines (2023-11-25)
Stefania Braidotti, Debora Curci, Daniele Zampieri, Cesare Covino, Davide Zanon, Natalia Maximova, Roberto Sala
ABSTRACT
Oligodendroglioma (OG) is a brain tumor that contributes to <1% of brain tumor diagnoses in the pediatric population. Unfortunately, pediatric OG remains without definitive molecular characteristics to aid in diagnosis, and little is known about the tumor microenvironment. Tumor cells' metabolism and proliferation rate are generally higher than those of healthy cells, so their iron demand is also significantly higher. This consideration underlines the great importance of iron for tumor development and progression. In this context, this study aims to evaluate the effect of iron in a cellular in vitro model of human oligodendroglioma brain tumor. Cell morphology, the effect of siderotic medium on cell growth, iron uptake, and the expression of iron-metabolism-related genes were evaluated via optic microscopy, ICP-MS, confocal microscopy, and real-time PCR, respectively. This study underlines the great importance of iron for tumor development and progression and also the possibility of reducing the available iron concentration to determine an antiproliferative effect on OG. Therefore, every attempt can be promising to defeat OG for which there are currently no long-term curative therapies.
Sigma-Aldrich
HOG Human Oligodendroglioma Cell Line, HOG human oligodendroglioma cell line is a useful model to study the role of oligodendrocytes in neurodegenerative diseases.