Merck
  • The Feline Immunodeficiency Virus Envelope Signal Peptide Is a Tetherin Antagonizing Protein.
The Feline Immunodeficiency Virus Envelope Signal Peptide Is a Tetherin Antagonizing Protein.
mBio (2023-03-18)
James H Morrison, Eric M Poeschla
ABSTRACT
Signal peptides are N-terminal peptides, generally less than 30 amino acids in length, that direct translocation of proteins into the endoplasmic reticulum and secretory pathway. The envelope glycoprotein (Env) of the nonprimate lentivirus feline immunodeficiency virus (FIV) contains the longest signal peptide of all eukaryotic, prokaryotic, and viral proteins (175 amino acids), yet the reason is unknown. Tetherin is a dual membrane-anchored host protein that inhibits the release of enveloped viruses from cells. Primate lentiviruses have evolved three antagonists: the small accessory proteins Vpu and Nef, and in the case of HIV-2, Env. Here, we identify the FIV Env signal peptide (Fsp) as the FIV tetherin antagonist. A short deletion in the central portion of Fsp had no effect on viral replication in the absence of tetherin, but severely impaired virion budding in its presence. Fsp is necessary and sufficient, acting as an autonomous accessory protein with the rest of Env dispensable. In contrast to primate lentivirus tetherin antagonists, its mechanism is to stringently block the incorporation of this restriction factor into viral particles rather than by degrading it or downregulating it from the plasma membrane. IMPORTANCE The study of species- and virus-specific differences in restriction factors and their antagonists has been central to deciphering the nature of these key host defenses. FIV is an AIDS-causing lentivirus that has achieved pandemic spread in the domestic cat. We now identify its tetherin antagonist as the signal sequence of the Envelope glycoprotein, thus identifying the fourth lentiviral anti-tetherin protein and the first new lentiviral accessory protein in decades. Fsp is necessary and sufficient and functions by stringently blocking particle incorporation of tetherin, which differs from the degradation or surface downregulation mechanisms used by primate lentiviruses. Fsp also is a novel example of signal peptide dual function, being both a restriction factor antagonist and a mediator of protein translocation into the endoplasmic reticulum.