Merck
  • Aberrant Expression of Solute Carrier Family 35 Member A2 Correlates With Tumor Progression in Breast Cancer.
Aberrant Expression of Solute Carrier Family 35 Member A2 Correlates With Tumor Progression in Breast Cancer.
In vivo (Athens, Greece) (2023-01-03)
Chien-Liang Liu, Shih-Ping Cheng, Wen-Chien Huang, Ming-Jen Chen, Chi-Hsin Lin, Shan-Na Chen, Yuan-Ching Chang
ABSTRACT
A recent study suggested that solute carrier family 35 member A2 (SLC35A2) is related to poor prognosis in patients with breast cancer. SLC35A2 transports uridine diphosphate-galactose from the cytosol to the lumen of the endoplasmic reticulum and Golgi. Immunohistochemical expression of SLC35A2 was evaluated using tissue microarrays. Cell growth, migration, and invasion of breast cancer cells were examined following loss- and gain-of-expression of SLC35A2. Normal breast tissue exhibited SLC35A2 immunoreactivity in the nucleus. A progressive increase in cytoplasmic expression from in situ carcinoma to invasive carcinoma was observed. There was a correlation between cytoplasmic SLC35A2 expression and breast cancer stage (p<0.001). MDA-MB-468 and MCF-7 cells transfected with SLC35A2 shRNA had unchanged cell viability but significantly reduced cell migration and invasion. In contrast, MDA-MB-231 and HCC1806 cells transfected with the SLC35A2 expression vector showed increased migration. Breast cancer progression is accompanied by differential expression patterns of SLC35A2. The migratory or invasive capacity of breast cancer cells is associated with SLC35A2 expression.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2