Merck
Mechanism of Ca2+ transport by ferroportin.
eLife (2023-01-18)
Jiemin Shen, Azaan Saalim Wilbon, Ming Zhou, Yaping Pan
ABSTRACT
Ferroportin (Fpn) is a transporter that releases ferrous ion (Fe2+) from cells and is important for homeostasis of iron in circulation. Export of one Fe2+ by Fpn is coupled to import of two H+ to maintain charge balance. Here, we show that human Fpn (HsFpn) binds to and mediates Ca2+ transport. We determine the structure of Ca2+-bound HsFpn and identify a single Ca2+ binding site distinct from the Fe2+ binding sites. Further studies validate the Ca2+ binding site and show that Ca2+ transport is not coupled to transport of another ion. In addition, Ca2+ transport is significantly inhibited in the presence of Fe2+ but not vice versa. Function of Fpn as a Ca2+ uniporter may allow regulation of iron homeostasis by Ca2+.
Sigma-Aldrich
Hepcidin-25 (human) trifluoroacetate salt, ≥95% (HPLC)