Merck
Microbiology and Preclinical Review of Omadacycline.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2019-08-02)
James A Karlowsky, Judith Steenbergen, George G Zhanel
ABSTRACT
Omadacycline is a novel aminomethylcycline antimicrobial and semisynthetic derivative of tetracycline. In vitro, omadacycline displays potent activity against gram-positive and many gram-negative bacteria, including methicillin-resistant Staphylococcus aureus, Streptococcus pneumoniae, β-hemolytic streptococci, vancomycin-resistant Enterococcus, and Enterobacteriaceae. Omadacycline is also active against atypical and anaerobic pathogens, including Legionella pneumophila, Mycoplasma spp., Ureaplasma spp., Bacteroides spp., and Clostridioides difficile. This review outlines the microbiology and preclinical studies of omadacycline, including its mechanism of action; spectrum of activity; protein binding; activity in the presence of surfactant, serum, normal, and pH-adjusted urine, or bacterial biofilms; postantibiotic effect; pharmacodynamic properties; and in vitro and in vivo efficacy. The results of in vitro and in vivo animal studies support the observations made in phase III clinical trials and the clinical development of omadacycline.