- Heparin inhibits mitogen-activated protein kinase-dependent and -independent c-fos induction in mesangial cells.
Heparin inhibits mitogen-activated protein kinase-dependent and -independent c-fos induction in mesangial cells.
Heparin suppresses mitogenic responses in renal mesangial cells, and when quiescent mesangial cells are stimulated with serum, heparin blocks the induction of c-fos seen at 15 min. Because heparin is taken up by cells over a much longer time course, we addressed mechanisms whereby extracellular heparin might suppress c-fos induction at such early times. Quiescent cells were treated with serum, 12-O-tetradecanoylphorbol-13-acetate, or low concentrations of Ca2+ ionophores that produced increases in intracellular Ca2+ concentration ([Ca2+]i) in the physiological range. Each treatment caused an increase in c-fos mRNA, but they did so by different mechanisms. Serum activated mitogen-activated protein kinase (MAPK) and increased [Ca2+]i without affecting protein kinase C. Activation of protein kinase C with phorbol ester activated MAPK without much effect on [Ca2+]i. Ionophores increased [Ca2+]i without affecting basal levels of protein kinase C or MAPK. Heparin (1 microg/ml) suppressed the induction of c-fos initiated by all three treatments. It did not affect the activity of protein kinase C, but inhibited activation of MAPK by either serum or phorbol ester, suggesting a common site of action at or below the probable convergence of the induced signals at Ras/Raf-1 activation. Heparin also inhibited the serum-stimulated entry of extracellular Ca2+ to the same extent as verapamil, consistent with the ability of verapamil to block L-type Ca2+ channels and the known presence of these channels in mesangial cells. However, this effect does not appear to be related to heparin's ability to inhibit induction of c-fos. First, verapamil had no effect on induction of c-fos by serum. Second, heparin had no effect on changes in [Ca2+]i achieved by ionophores. We conclude that heparin suppresses induction of c-fos in mesangial cells by blocking at least two different points in signal transduction cascades, one upstream of MAPK and the other independent of MAPK, but dependent on intracellular Ca2+.