Merck
  • Simultaneous determination of pesticides and mycotoxins in wine by direct injection and liquid chromatography-tandem mass spectrometry analysis.
Simultaneous determination of pesticides and mycotoxins in wine by direct injection and liquid chromatography-tandem mass spectrometry analysis.
Food chemistry (2019-06-04)
Jonatan V Dias, Maria da Graça P Nunes, Ionara R Pizzutti, Bárbara Reichert, Aliar A Jung, Carmem D Cardoso
ABSTRACT
A simple and rapid method for simultaneous determination of pesticides and mycotoxins in red wine is presented. Sample preparation approach, called direct injection, consists of a sequence of only three steps: centrifugation, dilution and filtration. The analysis of extracts were performed by UPLC-MS/MS for determination of pesticides and mycotoxins. The method was assessed for linearity, limits of detection and quantification, matrix effects, selectivity, accuracy and precision. For recovery experiments, mycotoxins were divided in two groups according to their sensitivity in the UPLC-MS/MS system. More than 80% of the mycotoxins were reliably quantified at the lowest spike level studied (1 μg kg-1 for group 1 and 50 μg kg-1 for group 2). From the 185 evaluated pesticides, 144 showed acceptable results at 10 μg kg-1, the lowest spiked level. Matrix effects were, in most of the cases, negative, and that was observed for both pesticides and mycotoxins.