Merck
  • Towards industrial production of microalgae without temperature control: The effect of diel temperature fluctuations on microalgal physiology.
Towards industrial production of microalgae without temperature control: The effect of diel temperature fluctuations on microalgal physiology.
Journal of biotechnology (2021-06-20)
Robin Barten, Yovita Djohan, Wendy Evers, Rene Wijffels, Maria Barbosa
ABSTRACT
Regions that offer high levels of sunlight are ideal to produce microalgae. However, as a result of high light intensities, the temperature in photobioreactors can reach temperatures up to 50 °C. Control of temperature is essential to avoid losses on biomass productivity but should be limited to a minimum to avoid high energy requirements for cooling. Our objective is to develop a production process in which cooling is not required. We studied the behaviour of thermotolerant microalgae Picochlorum sp. (BPE23) under four diel temperature regimes, with peak temperatures from 30 °C up to a maximum of 47.5 °C. The highest growth rate of 0.17 h-1 was obtained when applying a daytime peak temperature of 40 °C. Operating photobioreactors in tropical regions, with a maximal peak temperature of 40 °C, up from 30 °C, reduces microalgae production costs by 26.2 %, based on simulations with a pre-existing techno-economic model. Cell pigmentation was downregulated under increasingly stressful temperatures. The fatty acid composition of cell membranes was altered under increasing temperatures to contain shorter fatty acids with a higher level of saturation. Our findings show that the level of temperature control impacts the biomass yield and composition of the microalgae.
Sigma-Aldrich
Tripentadecanoin, ≥99%