Merck
  • Dexamethasone enhances the efficacy of atorvastatin in inhibiting excessively inflammation-induced abnormal angiogenesis by regulating macrophages.
Dexamethasone enhances the efficacy of atorvastatin in inhibiting excessively inflammation-induced abnormal angiogenesis by regulating macrophages.
Journal of neuroinflammation (2021-09-17)
Zhitao Gong, Daqiang Zhan, Meng Nie, Xiaochun Li, Chuang Gao, Xuanhui Liu, Tangtang Xiang, Jiangyuan Yuan, Weiwei Jiang, Jinhao Huang, Wei Quan, Dong Wang, Ye Tian, Hengjie Yuan, Jianning Zhang, Rongcai Jiang
ABSTRACT
We have recently showed that atorvastatin (ATO) combined with low dose of dexamethasone (DEX) was more efficacious in treating patients with chronic subdural haematoma (CSDH) than ATO monotherapy. This study was designed to investigate the underlying mechanisms of the improved efficacy of this combined therapy. Mass spectrometry was performed to quantitatively detect drugs in haematoma fluids and serum samples from CSDH patients and also in cultured macrophages after treatment with either ATO alone or in combination with DEX. The differentiation and apoptosis of macrophages were evaluated using flow cytometry. The expression of cytokines, chemokines and angiogenesis-related proteins was evaluated using proteome profile arrays, immunoblots and ELISA, respectively. ATO was detected in haematoma fluids and serum samples, whose levels were increased significantly in samples collected from patients treated with both ATO and DEX. ATO was also increased in cultured macrophages treated with ATO and DEX. The numbers of M1-polarized macrophages were higher than the M2 phenotype in the haematoma fluids of patients. Cultured macrophages treated with ATO and DEX had reduced numbers of M1-polarized macrophages, increased numbers of M2-polarized macrophages as compared to monotherapies, and decreased rate of apoptosis induced by high-dose DEX. DEX enhanced the anti-inflammatory and anti-angiogenic activity of ATO by suppressing VEGFA and other inflammatory angiogenic factors. Consistent with the finding, patients responded well to the drug treatments had lower serum levels of VEGFA. We have shown for the first time that ATO given orally was detected in CSDH haematoma fluids. DEX enhances the anti-inflammatory and anti-angiogenic effects of ATO, primarily by increasing the presence of ATO in haematoma and macrophages and by regulating the functions of macrophages.
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein