Merck
  • Galectin-1 Expression Is Associated with the Response and Survival Following Preoperative Chemoradiotherapy in Locally Advanced Esophageal Squamous Cell Carcinoma.
Galectin-1 Expression Is Associated with the Response and Survival Following Preoperative Chemoradiotherapy in Locally Advanced Esophageal Squamous Cell Carcinoma.
Cancers (2021-07-03)
Shau-Hsuan Li, Yen-Hao Chen, Hung-I Lu, Chien-Ming Lo, Chao-Cheng Huang, Yu-Ming Wang, Eng-Yen Huang
ABSTRACT
The galectin-1 has been found to be involved in poor outcomes after treatment of a variety of cancers. To the best of our knowledge, however, the significance of galectin-1 expression in the sensitivity to chemoradiotherapy (CCRT) of patients with locally advanced esophageal squamous cell carcinoma (ESCC) remains unclear. Expression levels of galectin-1 were evaluated by immunohistochemistry and correlated with the treatment outcome in 93 patients with locally advanced ESCC who received preoperative CCRT between 1999 and 2012. Galectin-1 expression was significantly associated with the pathological complete response (pCR). The pCR rates were 36.1% and 13.0% (p = 0.01) in patients with low and high galectin-1 expression, respectively. Univariate analyses revealed that galectin-1 overexpression, clinical 7th American Joint Committee on Cancer (AJCC) stage III and a positive surgical margin were significant factors of worse overall survival and disease-free survival. In multivariate analyses, galectin-1 overexpression and a positive surgical margin represented the independent adverse prognosticators. Therefore, galectin-1 expression both affects the pCR and survival in patients with locally advanced ESCC receiving preoperative CCRT. Our results suggest that galectin-1 may be a potentially therapeutic target for patients with ESCC treated with preoperative CCRT.