Merck
  • Preparation and applications of monoclonal antibodies to different collagen types.
Preparation and applications of monoclonal antibodies to different collagen types.
Clinical biochemistry (1988-04-01)
R Mayne
ABSTRACT
Monoclonal antibodies have recently been developed against all of the major collagen types isolated from both human and other species. These antibodies have several potential advantages over polyclonal antibodies, and a brief survey will be made of the different antibodies that have so far been developed. In addition, various successful applications of these antibodies to biological investigations will be briefly discussed.