Merck
  • The Δ40p53 isoform inhibits p53-dependent eRNA transcription and enables regulation by signal-specific transcription factors during p53 activation.
The Δ40p53 isoform inhibits p53-dependent eRNA transcription and enables regulation by signal-specific transcription factors during p53 activation.
PLoS biology (2021-08-06)
Cecilia B Levandowski, Taylor Jones, Margaret Gruca, Sivapriya Ramamoorthy, Robin D Dowell, Dylan J Taatjes
ABSTRACT
The naturally occurring Δ40p53 isoform heterotetramerizes with wild-type p53 (WTp53) to regulate development, aging, and stress responses. How Δ40p53 alters WTp53 function remains enigmatic because their co-expression causes tetramer heterogeneity. We circumvented this issue with a well-tested strategy that expressed Δ40p53:WTp53 as a single transcript, ensuring a 2:2 tetramer stoichiometry. Human MCF10A cell lines expressing Δ40p53:WTp53, WTp53, or WTp53:WTp53 (as controls) from the native TP53 locus were examined with transcriptomics (precision nuclear run-on sequencing [PRO-seq] and RNA sequencing [RNA-seq]), metabolomics, and other methods. Δ40p53:WTp53 was transcriptionally active, and, although phenotypically similar to WTp53 under normal conditions, it failed to induce growth arrest upon Nutlin-induced p53 activation. This occurred via Δ40p53:WTp53-dependent inhibition of enhancer RNA (eRNA) transcription and subsequent failure to induce mRNA biogenesis, despite similar genomic occupancy to WTp53. A different stimulus (5-fluorouracil [5FU]) also showed Δ40p53:WTp53-specific changes in mRNA induction; however, other transcription factors (TFs; e.g., E2F2) could then drive the response, yielding similar outcomes vs. WTp53. Our results establish that Δ40p53 tempers WTp53 function to enable compensatory responses by other stimulus-specific TFs. Such modulation of WTp53 activity may be an essential physiological function for Δ40p53. Moreover, Δ40p53:WTp53 functional distinctions uncovered herein suggest an eRNA requirement for mRNA biogenesis and that human p53 evolved as a tetramer to support eRNA transcription.
Sigma-Aldrich
Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)