Merck
Characterization of thymine microcrystals by CARS and SHG microscopy.
Scientific reports (2020-10-15)
Andrej Dementjev, Danielis Rutkauskas, Ivan Polovy, Mindaugas Macernis, Darius Abramavicius, Leonas Valkunas, Galina Dovbeshko
ABSTRACT
Identification of chemically homologous microcrystals in a polycrystal sample is a big challenge and requires developing specific highly sensitive tools. Second harmonic (SHG) and coherent anti-Stokes Raman scattering (CARS) spectroscopy can be used to reveal arrangement of thymine molecules, one of the DNA bases, in microcrystalline sample. Strong dependence of CARS and SHG intensity on the orientation of the linear polarization of the excitation light allows to obtain high resolution images of thymine microcrystals by additionally utilizing the scanning microscopy technique. Experimental findings and theoretical interpretation of the results are compared. Presented experimental data together with quantum chemistry-based theoretical interpretation allowed us to determine the most probable organization of the thymine molecules.
Sigma-Aldrich
Thymine, ≥99%