Merck
  • Single-Cell Transcriptomics of Human Mesenchymal Stem Cells Reveal Age-Related Cellular Subpopulation Depletion and Impaired Regenerative Function.
Single-Cell Transcriptomics of Human Mesenchymal Stem Cells Reveal Age-Related Cellular Subpopulation Depletion and Impaired Regenerative Function.
Stem cells (Dayton, Ohio) (2018-11-10)
Sacha M L Khong, Ming Lee, Nina Kosaric, Danika M Khong, Yixiao Dong, Ursula Hopfner, Matthias M Aitzetmüller, Dominik Duscher, Richard Schäfer, Geoffrey C Gurtner
ABSTRACT
Although bone marrow-derived mesenchymal stem cells (BM-MSCs) are widely recognized as promising therapeutic agents, the age-related impacts on cellular function remain largely uncharacterized. In this study, we found that BM-MSCs from young donors healed wounds in a xenograft model faster compared with their aged counterparts (p < .001). Given this significant healing advantage, we then used single-cell transcriptomic analysis to provide potential molecular insights into these observations. We found that the young cells contained a higher proportion of cells characterized by a higher expression of genes involved in tissue regeneration. In addition, we identified a unique, quiescent subpopulation that was exclusively present in young donor cells. Together, these findings may explain a novel mechanism for the enhanced healing capacity of young stem cells and may have implications for autologous cell therapy in the extremes of age. Stem Cells 2019;37:240-246.
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, Alexa Fluor 488 conjugate, clone 235-1, from mouse, ALEXA FLUOR 488