Merck
  • Helicobacter pylori isolated from Iranian drinking water: vacA, cagA, iceA, oipA and babA2 genotype status and antimicrobial resistance properties.
Helicobacter pylori isolated from Iranian drinking water: vacA, cagA, iceA, oipA and babA2 genotype status and antimicrobial resistance properties.
FEBS open bio (2016-07-16)
Reza Ranjbar, Faham Khamesipour, Nematollah Jonaidi-Jafari, Ebrahim Rahimi
ABSTRACT
Despite the clinical importance of Helicobacter pylori in human gastric disorders, its exact route of transmission is still uncertain. Based on the contentious hypothesis and findings of previous investigations, water may play an important role in the transmission of H. pylori to humans. This study was carried out to investigate the vacA, cagA, oipA, iceA and babA2 genotype status and antimicrobial resistance properties of H. pylori strains isolated from the drinking water samples of four major provinces in Iran. A total of 400 drinking water samples were cultured and tested. H. pylori-positive strains were analyzed for the presence of various genotypes and antimicrobial resistance. Twelve of 400 (3%) water samples were positive for H. pylori. Samples from Isfahan province had the highest, while those from Shiraz had the lowest prevalence of H. pylori. The seasonal distribution was also determined, with the highest prevalence of bacteria in the summer season (7.36%). H. pylori strains harbored the highest levels of resistance against ampicillin (100%), erythromycin (75%), clarithromycin (75%), and trimethoprim (58.3%). The most commonly detected genotypes were vacAs1a (83.3%), vacAm1a (66.6%), vacAs2 (50%) and cagA (50%). The presence of similar genotypes in the H. pylori strains of drinking water and those of human clinical samples suggest that contaminated water maybe the sources of bacteria. Spiramycin and furazolidone are suggested for the treatment of cases of H. pylori infection.
Product Description
Roche
DNA Isolation Kit for Cells and Tissues, sufficient for 10 isolation(s)