Merck
  • The replacement of His(4) in angiotensin IV by conformationally constrained residues provides highly potent and selective analogues.
The replacement of His(4) in angiotensin IV by conformationally constrained residues provides highly potent and selective analogues.
Journal of medicinal chemistry (2009-09-18)
Aneta Lukaszuk, Heidi Demaegdt, Debby Feytens, Patrick Vanderheyden, Georges Vauquelin, Dirk Tourwé
ABSTRACT
The histidine residue in angiotensin IV was replaced by various conformationally constrained amino acids. The substitution of the His(4)-Pro(5) dipeptide sequence by the constrained Trp analogue Aia-Gly, in combination with beta(2)hVal substitution at the N-terminus, provided a new stable analogue H-(R)-beta(2)hVal-Tyr-Ile-Aia-Gly-Phe-OH (AL-40) that is a potent ligand for the Ang IV receptor IRAP and selective versus AP-N and the AT1 receptor.