Merck
  • Dicer and Drosha expression and response to Bevacizumab-based therapy in advanced colorectal cancer patients.
Dicer and Drosha expression and response to Bevacizumab-based therapy in advanced colorectal cancer patients.
European journal of cancer (Oxford, England : 1990) (2012-12-26)
Bruno Vincenzi, Alice Zoccoli, Gaia Schiavon, Michele Iuliani, Francesco Pantano, Emanuela Dell'aquila, Raffaele Ratta, Andrea Onetti Muda, Giuseppe Perrone, Chiara Brunelli, Pierpaolo Correale, Elisabetta Riva, Antonio Russo, Fotios Loupakis, Alfredo Falcone, Daniele Santini, Giuseppe Tonini
ABSTRACT
The miRNA-regulating enzymes Dicer and Drosha exhibit aberrant expression in several cancer types. Dicer and Drosha play a crucial role during the angiogenetic process in vitro and, for Dicer, in vivo. We aimed to investigate the potential role of Dicer and Drosha in predicting response to Bevacizumab-based therapy in advanced colorectal cancer (CRC) patients. Dicer and Drosha mRNA levels were analysed in formalin-fixed paraffin-embedded specimens from patients affected by advanced CRC treated with or without Bevacizumab-containing regimens (n=116 and n=50, respectively) and from patients with diverticulosis as control group (n=20). The experimental data were obtained using qRT-PCR, analysed comparing Dicer and Drosha expression levels in tumour samples versus normal mucosa and then compared to clinical outcome. The tumour samples from Bevacizumab-treated patients showed a significantly higher Drosha expression (P<.001) versus normal mucosa, while Dicer levels did not differ. Intriguingly, we found that low Dicer levels predicted a longer progression-free survival (PFS) (P<.0001) and overall survival (OS) (P=.009). In addition, low Dicer levels were associated with better response to Bevacizumab-based treatments versus high Dicer levels (1.7% complete responses and 53.4% partial responses versus 0% and 32.7%, respectively; P=.0067). Multivariate analysis identified three independent predictors of improved OS: high performance status (PS) (relative risk (RR) 1.45; P=.011), lower organs involvement (RR 0.79; P=.034) and low Dicer expression (RR 0.71; P=.008). Conversely, Drosha levels were not associated with prognosis and outcome associated with treatment. In non-Bevacizumab-treated patients, Dicer and Drosha expression did not correlate with outcome. These findings suggest that low Dicer mRNA levels seem to be independent predictors of favourable outcome and response in patients affected by advanced CRCs treated with Bevacizumab-based therapy.