Merck
  • miR-202 Suppresses Hepatocellular Carcinoma Progression via Downregulating BCL2 Expression.
miR-202 Suppresses Hepatocellular Carcinoma Progression via Downregulating BCL2 Expression.
Oncology research (2020-04-11)
Donghai Zhuang, Li Liang, Hongzhan Zhang, Xianguang Feng
ABSTRACT
miRNAs play an important role in progression of hepatocellular carcinoma (HCC). In this work, we assessed the function of miR-202 in human HCC and identified BCL2 as its target. We found miR-202 expression was found significantly downregulated, while BCL2 expression was markedly upregulated in HCC tissues and cell lines (HepG2, Hep3B, and HCCLM3). Both miR-202 and BCL2 were closely correlated with major vascular invasion and advanced TNM stage as well as overall survival of HCC patients. Overexpression of miR-202 significantly inhibited cell proliferation, induced apoptosis and cell cycle arrest at the G0/G1 phase, and prevented tumor formation in a xenograft nude mouse model. Further, miR-202 dramatically inhibited migration, invasion, and epithelialmesenchymal transition. miR-202 bound to the 3-untranslated region (3-UTR) of BCL2 mRNA and downregulated the expression level of BCL2 protein. Exogenous BCL2 overexpression weakened the inhibitory effects of miR-202, while inhibition of BCL2 enhanced the inhibitory effects of miR-202. In conclusion, miR-202 serves as a tumor suppressor in HCC progression by downregulating BCL2 expression, indicating miR-202 might be a potential target for HCC.