Merck
  • Design and evaluation of hydroxamate derivatives as metal-mediated inhibitors of a protein tyrosine kinase.
Design and evaluation of hydroxamate derivatives as metal-mediated inhibitors of a protein tyrosine kinase.
Journal of medicinal chemistry (2006-12-08)
Xianfeng Gu, Yuehao Wang, Anil Kumar, Guofeng Ye, Keykavous Parang, Gongqin Sun
ABSTRACT
Protein tyrosine kinases use two Mg(2+) ions as cofactors in catalysis, one as the ATP-Mg complex (M1) and the other as an essential activator (M2). The M2-binding site has high affinity for transition metal cations such as cobalt and zinc. Taking advantage of this high affinity, we examined hydroxamates as metal-mediated inhibitors against C-terminal Src kinase (Csk), a protein tyrosine kinase. Of a small group of amino acid hydroxamates, tyrosine and phenylalanine hydroxamates inhibited Csk activity only in the presence of Co(2+). Four classes of phenylalanine and tyrosine hydroxamate derivatives were synthesized and evaluated as metal-mediated inhibitors of Csk, leading to improved inhibition and a better understanding of the structure-activity relationships. This study suggests that hydroxamates may serve as a general scaffold for developing metal-mediated inhibitors against protein tyrosine kinases. To the best of our knowledge, this is the first report of designing metal-mediated inhibitors against a protein tyrosine kinase by targeting a metal binding site.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
SAFC
L-Tyrosine