Merck
  • An indispensable role for dynamin-related protein 1 in beige and brown adipogenesis.
An indispensable role for dynamin-related protein 1 in beige and brown adipogenesis.
Journal of cell science (2020-08-28)
Raja Gopal Reddy Mooli, Dhanunjay Mukhi, Zhonghe Chen, Nia Buckner, Sadeesh K Ramakrishnan
ABSTRACT
Emerging evidence indicates that proper mitochondrial dynamics are critical for adipocyte differentiation and functional thermogenic capacity. We found that the mitochondrial fission protein dynamin-related protein 1 (DRP1, also known as DNML1) is highly expressed in brown adipose tissue compared to expression in white adipose tissue, and these expression levels increase during brown adipocyte differentiation. Our results reveal that the inhibition of DRP1 using mdivi-1 mitigates beige adipocyte differentiation and differentiation-associated mitochondrial biogenesis. We found that DRP1 is essential for the induction of the early-phase beige adipogenic transcriptional program. Intriguingly, inhibition of DRP1 is dispensable following the induction of beige adipogenesis and adipogenesis-associated mitochondrial biogenesis. Altogether, we demonstrate that DRP1 in preadipocytes plays an essential role in beige and brown adipogenesis.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
MISSION® esiRNA, targeting human DNM1L