Merck
  • Cytotoxic mechanisms of primin, a natural quinone isolated from Eugenia hiemalis, on hematological cancer cell lines.
Cytotoxic mechanisms of primin, a natural quinone isolated from Eugenia hiemalis, on hematological cancer cell lines.
Anti-cancer drugs (2020-07-09)
Alisson Bigolin, Mariana F Maioral, Natália M Stefanes, Gabriele A Zatelli, Ana C Philippus, Miriam de B Falkenberg, Maria C Santos-Silva
ABSTRACT
Considering the high morbidity and mortality rates associated with hematological malignancies and the frequent development of drug resistance by these diseases, the search for new cytotoxic agents is an urgent necessity. The new compounds should present higher efficiency and specificity in inducing tumor cell death, be easily administered and have little or negligible adverse effects. Quinones have been reported in the literature by their several pharmacological properties, including antitumor activity, thus, the aim of this study was to investigate the cytotoxic effect of primin, a natural quinone, on hematological malignancies cell lines. Primin was highly cytotoxic against the three cell lines included in this study (K562, Jurkat and MM.1S) in a concentration- and time-dependent manner, as demonstrated by the MTT method. The compound triggered an apoptotic-like cell death, as observed by ethidium bromide/acridine orange staining, DNA fragmentation and phosphatidylserine exposure after labeling with Annexin V. Both intrinsic and extrinsic apoptosis are involved in cell death induced by primin, as well as the modulation of cell proliferation marker KI-67. The activation of intrinsic apoptosis appears to be related to a decreased Bcl-2 expression and increased Bax expression. While the increase in FasR expression signals activate extrinsic apoptosis. The results suggest that primin is a promising natural molecule that could be used in hematological malignancies therapy or as prototypes for the development of new chemotherapics.