Merck
  • miR-182 prevented ototoxic deafness induced by co-administration of kanamycin and furosemide in rats.
miR-182 prevented ototoxic deafness induced by co-administration of kanamycin and furosemide in rats.
Neuroscience letters (2020-02-28)
Jun Chen, Zhenzhen Liu, Hui Yan, Wei Xing, Wenjuan Mi, Renfeng Wang, Wei Li, Fuquan Chen, Jianhua Qiu, Dingjun Zha
ABSTRACT
Ototoxic drugs may induce auditory sensory hair cell loss and permanent deafness; however, there is still no effective treatments or prevention strategies for this side effect. A recent study found that microRNA182 (miR-182) protected cochlear hair cells from ototoxic drug-induced apoptosis in vitro. However, it remains unclear whether miR-182 can protect drug-induced deafness in vivo. In this study, we overexpressed cochlear miR-182 in Sprague-Dawley rats by trans-round window niche delivery of miR-182 mimics. The rats subsequently received intraperitoneal injections of kanamycin and furosemide to induce acute cochlear outer hair cell death and permanent deafness. Auditory brainstem response tests showed that miR-182 attenuated permanent threshold shifts. Consistent with this result, miR-182 reduced the loss of outer hair cells and missing stereocilia. miR-182 treatment also increased the level of phosphoinositide-3 kinase regulatory subunit p85α in the outer hair cells after co-administration of kanamycin and furosemide. Our findings suggest that miR-182 has powerful protective potential against ototoxic drug-induced acute auditory sensory hair cell loss and permanent deafness.