Merck
  • Pymetrozine inhibits reproductive behavior of brown planthopper Nilaparvata lugens and fruit fly Drosophila melanogaster.
Pymetrozine inhibits reproductive behavior of brown planthopper Nilaparvata lugens and fruit fly Drosophila melanogaster.
Pesticide biochemistry and physiology (2020-05-04)
Li-Xiang Wang, Yan-Chao Zhang, Sha Tao, Di Guo, Yan Zhang, Ya-Long Jia, Shuai Zhang, Chen Zheng, Dilawar Khan, Cong-Fen Gao, Shun-Fan Wu
ABSTRACT
Pymetrozine is a promising chemical used to control brown planthopper, which developed resistance to imidacloprid and buprofezin in the past decade. Field efficacy indicates that pymetrozine can reduce the number of offsprings of brown planthopper, but the specific physiological mechanism is unknown. In this study, we systematically described the mating process of brown planthopper including 8 steps (abdominal vibration, following, positioning, wing extension, attempted copulation, copulation, terminated copulation and leaving) and explored the optimal mating time after adult eclosion (3-5 days) and observation time (30 mins). Also, behavioral data showed that pymetrozine can affect the mating behavior and female fecundity of brown planthopper and fruit fly. As one of the target genes for pymetrozine, Nanchung (Nan), the nan36a mutant male courtship index, female receptivity and the number of offsprings were significantly decreased. Behavioral defects in nan36a mutant flies can be rescued by expressed NlNan. Our results indicated that Nan plays essential roles in the mating behavior and female fecundity. These findings provide useful information for demonstrating that pymetrozine effectively reduce the reproduction of brown planthopper and contribute to our understanding of reproductive strategies controlled by pymetrozine in insects.
Product Description
Supelco
Pymetrozin, PESTANAL®, analytical standard