Merck
  • Aryl Hydrocarbon Receptor Activation and Tissue Factor Induction by Fluid Shear Stress and Indoxyl Sulfate in Endothelial Cells.
Aryl Hydrocarbon Receptor Activation and Tissue Factor Induction by Fluid Shear Stress and Indoxyl Sulfate in Endothelial Cells.
International journal of molecular sciences (2020-04-05)
Guillaume Lano, Manon Laforêt, Clarissa Von Kotze, Justine Perrin, Tawfik Addi, Philippe Brunet, Stéphane Poitevin, Stéphane Burtey, Laetitia Dou
ABSTRACT
Endogenous agonists of the transcription factor aryl hydrocarbon receptor (AHR) such as the indolic uremic toxin, indoxyl sulfate (IS), accumulate in patients with chronic kidney disease. AHR activation by indolic toxins has prothrombotic effects on the endothelium, especially via tissue factor (TF) induction. In contrast, physiological AHR activation by laminar shear stress (SS) is atheroprotective. We studied the activation of AHR and the regulation of TF by IS in cultured human umbilical vein endothelial cells subjected to laminar fluid SS (5 dynes/cm2). SS and IS markedly increased the expression of AHR target genes PTGS2 (encoding for COX2), AHRR, CYP1A1, and CYP1B1, as well as F3 (encoding for TF), in an AHR-dependent way. IS amplified SS-induced TF mRNA and protein expression and upregulation of AHR target genes. Interestingly, tyrosine kinase inhibition by genistein decreased SS- but not IS-induced TF expression. Finally, the increase in TF expression induced by laminar SS was not associated with increased TF activity. In contrast, IS increased TF activity, even under antithrombotic SS conditions. In conclusion, IS and SS induce AHR activation and AHR-dependent TF upregulation by different mechanisms. Impairment of the antithrombotic properties of shear stressed endothelium by toxic AHR agonists could favor cardiovascular diseases in CKD.