Merck
  • Stat3 activation induces insulin resistance via a muscle-specific E3 ubiquitin ligase Fbxo40.
Stat3 activation induces insulin resistance via a muscle-specific E3 ubiquitin ligase Fbxo40.
American journal of physiology. Endocrinology and metabolism (2020-02-27)
Liping Zhang, Zihong Chen, Ying Wang, David J Tweardy, William E Mitch
ABSTRACT
Cellular mechanisms causing insulin resistance (IR) in chronic kidney disease (CKD) are poorly understood. One potential mechanism is that CKD-induced inflammation activates the signal transducer and activator of transcription 3 (Stat3) in muscle. We uncovered increased p-Stat3 in muscles of mice with CKD or mice fed high-fat diet (HFD). Activated Stat3 stimulates the expression of Fbxo40, a muscle-specific E3 ubiquitin ligase that stimulates ubiquitin conjugation leading to degradation of insulin receptor substrate 1 (IRS1). Evidence that Stat3 activates Fbxo40 includes 1) potential Stat3 binding sites in Fbxo40 promoters; 2) Stat3 binding to the Fbxo40 promoter; and 3) constitutively active Stat3 stimulating both Fbxo40 expression and its promoter activity. We found that IL-6 activates Stat3 in myotubes, increasing Fbxo40 expression with reduced IRS1 and p-Akt. Knockdown Fbxo40 using siRNA from myotubes results in higher levels of IRS1 and p-Akt despite the presence of IL-6. We treated mice with a small-molecule inhibitor of Stat3 (TTI-101) and found improved glucose tolerance and insulin signaling in skeletal muscles of mice with CKD or fed an HFD. Finally, we uncovered improved glucose tolerance in mice with muscle-specific Stat3 KO versus results in Stat3f/f mice in response to the HFD. Thus Stat3 activation in muscle increases IR in mice. Inhibition of Stat3 by TTI-101 could be developed into clinical strategies to improve muscle insulin signaling in inflammation and other catabolic diseases.