Merck
  • A Multisite-Binding Switchable Fluorescent Probe for Monitoring Mitochondrial ATP Level Fluctuation in Live Cells.
A Multisite-Binding Switchable Fluorescent Probe for Monitoring Mitochondrial ATP Level Fluctuation in Live Cells.
Angewandte Chemie (International ed. in English) (2015-12-18)
Lu Wang, Lin Yuan, Xian Zeng, Juanjuan Peng, Yong Ni, Jun Cheng Er, Wang Xu, Bikram Keshari Agrawalla, Dongdong Su, Beomsue Kim, Young-Tae Chang
ABSTRACT
Adenosine triphosphate (ATP), commonly produced in mitochondria, is required by almost all the living organisms; thus fluorescent probes for monitoring mitochondrial ATP levels fluctuation are essential and highly desired. Herein, we report a multisite-binding switchable fluorescent probe, ATP-Red 1, which selectively and rapidly responds to intracellular concentrations of ATP. Live-cell imaging indicated that ATP-Red 1 mainly localized to mitochondria with good biocompatibility and membrane penetration. In particular, with the help of ATP-Red 1, we successfully observed not only the decreased mitochondrial ATP levels in the presence of KCN and starvation state, but also the increased mitochondrial ATP levels in the early stage of cell apoptosis. These results indicate that ATP-Red 1 is a useful tool for investigating ATP-relevant biological processes.
Sigma-Aldrich
BioTracker ATP-Red Live Cell Dye, Live cell imaging dye for cellular adenosine triphosphate (ATP) localized to mitochondria used to detect metabolic activity, cell proliferation and overall cell health.