Merck
  • Palladium-catalyzed esterification of aryl halides using aryl formates without the use of external carbon monoxide.
Palladium-catalyzed esterification of aryl halides using aryl formates without the use of external carbon monoxide.
Chemical communications (Cambridge, England) (2012-07-10)
Tetsuaki Fujihara, Tomoya Hosoki, Yuko Katafuchi, Tomohiro Iwai, Jun Terao, Yasushi Tsuji
ABSTRACT
Aryl formates are efficient carbon monoxide sources in palladium-catalyzed esterification of aryl halides. The carbonylation readily proceeds at ambient pressure without the use of external carbon monoxide to afford the corresponding esters in high yields.
Sigma-Aldrich
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), 95%