Merck
  • Cellular Turnover: A Potential Metabolic Rate-Driven Mechanism to Mitigate Accumulation of DNA Damage.
Cellular Turnover: A Potential Metabolic Rate-Driven Mechanism to Mitigate Accumulation of DNA Damage.
Physiological and biochemical zoology : PBZ (2020-02-06)
Stanisław Bury, Agnieszka Cierniak, Joanna Jakóbik, Edyta T Sadowska, Mariusz Cichoń, Ulf Bauchinger
ABSTRACT
Oxidative stress, the imbalance of reactive oxygen species and antioxidant capacity, may cause damage to biomolecules pivotal for cellular processes (e.g., DNA). This may impair physiological performance and, therefore, drive life-history variation and aging rate. Because aerobic metabolism is supposed to be the main source of such oxidative risk, the rate of oxygen consumption should be positively associated with the level of damage and/or antioxidants. Empirical support for such relationships remains unclear, and recent considerations suggest even a negative relationship between metabolic rate and oxidative stress. We investigated the relationship between standard metabolic rate (SMR), antioxidants, and damage in blood plasma and erythrocytes for 35 grass snakes (Natrix natrix). Reactive oxygen metabolites (dROMs) and nonenzymatic antioxidants were assessed in plasma, while two measures of DNA damage and the capacity to neutralize H2O2 were measured in erythrocytes. Plasma antioxidants showed no correlation to SMR, and the level of dROMs was positively related to SMR. A negative relationship between antioxidant capacity and SMR was found in erythrocytes, but no association of SMR with either measure of DNA damage was detected. No increase in DNA damage, despite lower antioxidant capacity at high SMR, indicates an upregulation in other defense mechanisms (e.g., damage repair and/or removal). Indeed, we observed a higher frequency of immature red blood cells in individuals with higher SMR, which indicates that highly metabolic individuals had increased erythrocyte turnover, a mechanism of damage removal. Such DNA protection through upregulated cellular turnover might explain the negligible senescence observed in some ectotherm taxa.
Product Description
Supelco
N-Methyl-N-trimethylsilyltrifluoroacetamide activated III, derivatization grade (GC derivatization), LiChropur, activated with imidazole