Merck
  • The growing prevalence of Nosema ceranae in honey bees in Spain, an emerging problem for the last decade.
The growing prevalence of Nosema ceranae in honey bees in Spain, an emerging problem for the last decade.
Research in veterinary science (2011-09-13)
Cristina Botías, Raquel Martín-Hernández, Encarna Garrido-Bailón, Amelia González-Porto, Amparo Martínez-Salvador, Pilar De la Rúa, Aránzazu Meana, Mariano Higes
ABSTRACT
Microsporidiosis caused by infection with Nosema apis or Nosema ceranae has become one of the most widespread diseases of honey bees and can cause important economic losses for beekeepers. Honey can be contaminated by spores of both species and it has been reported as a suitable matrix to study the field prevalence of other honey bee sporulated pathogens. Historical honey sample collections from the CAR laboratory (Centro Apícola Regional) were analyzed by PCR to identify the earliest instance of emergence, and to determine whether the presence of Nosema spp. in honey was linked to the spread of these microsporidia in honey bee apiaries. A total of 240 frozen honey samples were analyzed by PCR and the results compared with rates of Nosema spp. infection in worker bee samples from different years and geographical areas. The presence of Nosema spp. in hive-stored honey from naturally infected honey bee colonies (from an experimental apiary) was also monitored, and although collected honey bees resulted in a more suitable sample to study the presence of microsporidian parasites in the colonies, a high probability of finding Nosema spp. in their hive-stored honey was observed. The first honey sample in which N. ceranae was detected dates back to the year 2000. In subsequent years, the number of samples containing N. ceranae tended to increase, as did the detection of Nosema spp. in adult worker bees. The presence of N. ceranae as early as 2000, long before generalized bee depopulation and colony losses in 2004 may be consistent with a long incubation period for nosemosis type C or related with other unknown factors. The current prevalence of nosemosis, primarily due to N. ceranae, has reached epidemic levels in Spain as confirmed by the analysis of worker honey bees and commercial honey.
Product Description
Roche
FastStart PCR Master, sufficient for ≤100 reactions (04710436001), sufficient for ≤400 reactions (04710444001), sufficient for ≤2,000 reactions (04710452001)