Merck
  • MicroRNA-431 serves as a tumor inhibitor in breast cancer through targeting FGF9.
MicroRNA-431 serves as a tumor inhibitor in breast cancer through targeting FGF9.
Oncology letters (2020-01-04)
Wei Wang, Yan Dong, Xiaoyan Li, Yingying Pan, Jiexin Du, Daotong Liu
ABSTRACT
Breast cancer has become an important public health problem. Moreover, the functions of microRNA-431 (miR-431) have been detected in human cancers other than breast cancer. Hence, we investigated the role of miR-431 in progression of breast cancer. RT-qPCR and Western blot analysis were performed to assess expression of miR-431 and genes. The regulatory mechanism of miR-431 was investigated using MTT, Transwell and luciferase reporter assay. Decreased miR-431 expression was identified in breast cancer, which was related to aggressive behavior. Furthermore, miR-431 restrained cell proliferation, metastasis and EMT in breast cancer. miR-431 induced apoptosis through enhancing Bax expression. In addition, miR-431 was found to directly target FGF9. Moreover, upregulation of FGF9 impaired the anti-tumor effect of miR-431 in breast cancer. miR-431 restrained cell viability and metastasis in breast cancer through targeting FGF9, indicating that miR-431 serves as a tumor inhibitor in breast cancer.