Merck
[Polyclonal antibodies against recombinant dengue virus NS3 protein].
Biomedica : revista del Instituto Nacional de Salud (2017-05-21)
Liliana Morales, Myriam L Velandia, María Angélica Calderon, Jaime E Castellanos, Jacqueline Chaparro-Olaya
ABSTRACT
Dengue is a disease caused by one of four serotypes of the dengue virus (DENV) and is endemic in approximately 130 countries. The incidence of dengue has increased dramatically in recent decades, as well as the frequency and magnitude of outbreaks. Despite all efforts, there are no prophylactic or therapeutic treatments for the disease. Accordingly, research on the processes governing the DENV infection cycle is essential to develop vaccines or antiviral therapies. One of themost attractive DENV molecules to investigate is nonstructural protein 3 (NS3), which is essential for viral replication and a major immune target for infection. To produce antibodies to support future studies on NS3 and its cellular interactions with other proteins. Two recombinant proteins of the helicase domain of DENV NS3 serotype 2 were expressed, and used to immunize mice and produce polyclonal antibodies. The antibodies produced were useful in Western blot and immunofluorescence tests. We report an NS3 antibody that immunoprecipitates the viral protein and detects it in Western blot with no need to over-express it or use cell extracts with metabolic radiolabeling. The recombinant proteins expressed and the antibodies produced constitute valuable tools for studying DENV infectious processes involving NS3 and for evaluating tests designed to interfere with its functions.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-St. Louis Encephalitis Antibody, MSI-7, clone 6B6C-1, clone 6b6c-1, Chemicon®, from mouse