Merck
  • Regional differences in Ca2+ entry along the proximal-middle-distal muscle axis during eccentric contractions in rat skeletal muscle.
Regional differences in Ca2+ entry along the proximal-middle-distal muscle axis during eccentric contractions in rat skeletal muscle.
Journal of applied physiology (Bethesda, Md. : 1985) (2019-08-02)
Ayaka Tabuchi, Hiroaki Eshima, Yoshinori Tanaka, Shunsuke Nogami, Naoki Inoue, Mizuki Sudo, Hidetaka Okada, David C Poole, Yutaka Kano
ABSTRACT
Eccentric (ECC) contraction-induced muscle damage is associated with calcium ion (Ca2+) influx from the extracellular milieu through stretch-activated channels. It remains unknown whether Ca2+ influx consequent to repetitive ECC contractions is nonuniform across different muscle regions. We tested the hypothesis that there are regional differences in Ca2+ entry along the proximal-middle-distal muscle axis. Tibialis anterior (TA) muscles of adult male Wistar rats were exposed by reflecting the overlying skin and fasciae and ECC contractions evoked by peroneal nerve stimulation paired with simultaneous ankle extension (50 times/set, 2 protocols: 1 set and 10 sets). During ECC in the proximal, middle, and distal TA, we determined 1) muscle fiber extension by high-speed camera (200 frames/s) and 2) Ca2+ accumulation by in vivo bioimaging (Ca2+-sensitive probe Fura-2-acetoxymethyl ester). Muscle fiber extension from resting was significantly different among regions (i.e., proximal, 4.0%: < middle, 11.2%: < distal, 17.0%; ECC phase length at 500th contraction). Intracellular Ca2+ accumulation after 1 set of ECC was higher in the distal (1.46 ± 0.04, P < 0.05) than the proximal (1.27 ± 0.04) or middle (1.26 ± 0.05) regions. However, this regional Ca2+ accumulation difference disappeared by 32.5 min after the 1 set protocol when the muscle was quiescent and by contraction set 5 for the 10-set protocol. The initial preferential ECC-induced Ca2+ accumulation observed distally was associated spatially with the greater muscle extension compared with that of the proximal and middle regions. Disappearance of the regional Ca2+ accumulation disparity in quiescent and ECC-contracting muscle might be explained, in part, by axial Ca2+ propagation and account for the uniformity of muscle damage across regions evident 3 days post-ECC.NEW & NOTEWORTHY After 1 set of 50 eccentric (ECC) contractions in the anterior tibialis muscle, intracellular Ca2+ ([Ca2+]i) accumulation evinces substantial regional heterogeneity that is spatially coherent with muscle length changes (i.e., distal [Ca2+]i > middle, proximal). However, irrespective of whether 50 or 500 ECC contractions are performed, this heterogeneity is subsequently abolished, at least in part, by axial intracellular Ca2+ propagation. This Ca2+ homogenization across regions is consistent with the absence of any interregional difference in muscle damage 3 days post-ECC.
Sigma-Aldrich
Monoclonal Anti-μ-Calpain, Large Subunit antibody produced in mouse, clone 15C10, purified immunoglobulin