- Leuhistin, a new inhibitor of aminopeptidase M, produced by Bacillus laterosporus BMI156-14F1. I. Taxonomy, production, isolation, physico-chemical properties and biological activities.
Leuhistin, a new inhibitor of aminopeptidase M, produced by Bacillus laterosporus BMI156-14F1. I. Taxonomy, production, isolation, physico-chemical properties and biological activities.
The Journal of antibiotics (1991-06-01)
T Aoyagi, S Yoshida, N Matsuda, T Ikeda, M Hamada, T Takeuchi
PMID1676997
ABSTRACT
Leuhistin has been isolated from the culture broth of Bacillus laterosporus BMI156-14F1 as part of a program designed to find microorganism-produced inhibitors of aminopeptidase M (AP-M). It was purified by use of column chromatography on Sepabeads SP206, Amberlite IRC-50, MCI gel CHP-20P and Sephadex G-10 and then isolated as colorless needles. Leuhistin inhibits AP-M strongly and it also inhibits AP-A and AP-B weakly. It is competitive with the substrate, and the inhibition constant (Ki) was 2.3 x 10(-7) M.