Merck
  • Biochemical characteristics of a nitroreductase with diverse substrate specificity from Streptomyces mirabilis DUT001.
Biochemical characteristics of a nitroreductase with diverse substrate specificity from Streptomyces mirabilis DUT001.
Biotechnology and applied biochemistry (2018-09-20)
Jun Yang, Jing Bai, Mingbo Qu, Bo Xie, Qing Yang
ABSTRACT
A nitroreductase-encoded gene from an efficient nitro-reducing bacterium Streptomyces mirabilis DUT001, named snr, was cloned and heterogeneously expressed in Escherichia coli. The purified Streptomyces nitroreductase SNR was a homodimer with an apparent subunit molecular weight of 24 kDa and preferred NADH to NADPH as a cofactor. By enzyme incubation and isothermal calorimetry experiments, flavin mononucleotide (FMN) was found to be the preferred flavin cofactor; the binding process was exothermic and primarily enthalpy driven. The enzyme can reduce multiple nitro compounds and flavins, including antibacterial drug nitrofurazone, priority pollutants 2,4-dinitrotoluene and 2,4,6-trinitrotoluene, as well as key chemical intermediates 3-nitrophthalimide, 4-nitrophthalimide, and 4-nitro-1,8-naphthalic anhydride. Among the substrates tested, the highest activity of kcat(app) /Km(app) (0.234 μM-1  Sec-1 ) was observed for the reduction of FMN. Multiple sequence alignment revealed that the high FMN reduction activity of SNR may be due to the absence of a helix, constituting the entrance to the substrate pocket in other nitroreductases.