Merck
  • Metabolic Profiling and Cold-Starvation Stress Response of Oxygen-Tolerant Lactobacillus gasseri Strains Cultured in Batch Bioreactor.
Metabolic Profiling and Cold-Starvation Stress Response of Oxygen-Tolerant Lactobacillus gasseri Strains Cultured in Batch Bioreactor.
Microorganisms (2019-07-18)
Diamante Maresca, Francesca De Filippis, Alessandro Robertiello, Gianluigi Mauriello
ABSTRACT
Phenotypic and genotypic evidence indicates that many LAB strains can grow in presence of oxygen and can shift from fermentative to aerobic and/or respiratory metabolism. The aerobic and respiratory growth of several LAB species have been studied, allowing the selection of strains showing improved biomass production, long-term survival, and resistance under oxygen and stress conditions. The aim of this work was to observe the adaptation of two Lactobacillus gasseri strains, described in a previous work, to aerobic (air injection) and respiratory (air injection plus hemin and menaquionone) conditions obtained in a batch bioreactor. One strain showed the higher biomass production and oxygen consumption as well as the lower acidification in respiratory condition. Instead, the other one grew better in aerobic condition, even though the higher resistance to cold-starvation stress was registered in respiratory condition. In silico analysis revealed notable differences between AL3 and AL5 genomes and that of the type strain. This work contributes to understanding the adaptation response of lactobacilli to aerobic and respiratory metabolism. We demonstrated that the supposed activation of respiratory metabolism may provide several modifications to cell physiology. These features may be relevant in some technological and health-promoting applications, including starter and probiotic formulations.