Merck
  • Serum enterolactone concentrations are low in colon but not in rectal cancer patients.
Serum enterolactone concentrations are low in colon but not in rectal cancer patients.
Scientific reports (2019-08-03)
Anne Tuomisto, Natalja P Nørskov, Päivi Sirniö, Juha P Väyrynen, Shivaprakash J Mutt, Kai Klintrup, Jyrki Mäkelä, Knud Erik Bach Knudsen, Markus J Mäkinen, Karl-Heinz Herzig
ABSTRACT
The dietary lignan metabolite, enterolactone, has been suggested to have anti-cancer functions, and high serum enterolactone concentrations have been associated with decreased risk of breast and prostate cancers. We hypothesized that serum enterolactone concentrations as a marker of plant-based foods are associated with decreased risk in colorectal cancer (CRC). We measured serum enterolactone glucuronide and sulfate concentrations by liquid chromatography-tandem mass spectrometry in 115 CRC patients and 76 sex- and age-matched controls and analyzed the results with respect to tumor parameters, clinical parameters, and systemic inflammatory markers. Patients with colon cancer had significant lower serum enterolactone glucuronide and sulfate concentrations than controls (glucuronide: median 3.14 nM vs. 6.32 nM, P < 0.001; sulfate: median 0.13 nM vs. 0.17 nM, P = 0.002), whereas rectal cancer patients had similar enterolactone levels as controls (glucuronide: median 5.39 nM vs. 6.32 nM, P = 0.357; sulfate: median 0.19 nM vs. 0.17 nM, P = 0.452). High serum enterolactone concentrations were associated with low tumor grade, high serum creatinine levels, and concomitant diabetes. In summary, our results suggest that serum enterolactone concentrations are decreased in colon but not in rectal cancer. Further investigations are required to assess whether this reflects an altered lignan metabolism by the colon microbiome.