- Practical Microwave Synthesis of Carbazole Aldehydes for the Development of DNA-Binding Ligands.
Practical Microwave Synthesis of Carbazole Aldehydes for the Development of DNA-Binding Ligands.
Molecules (Basel, Switzerland) (2019-03-13)
Agata Głuszyńska, Bernard Juskowiak
PMID30857275
ABSTRACT
Microwave formylation of carbazole derivatives was investigated and 3-monoaldehydes were obtained in high yield. A potential DNA-binding ligand, 3-[(3-ethyl)-2-vinylbenzothiazolium]-9-N-ethyl carbazole iodide, was synthesized and characterized including spectral properties (UV-Vis absorption and fluorescence spectra). The binding selectivity and affinity of three carbazole ligands for double-stranded and G-quadruplex DNA structures were studied using a competitive dialysis method in sodium- and potassium-containing buffer solutions.