- Tn antigen promotes human colorectal cancer metastasis via H-Ras mediated epithelial-mesenchymal transition activation.
Tn antigen promotes human colorectal cancer metastasis via H-Ras mediated epithelial-mesenchymal transition activation.
Tn antigen is a truncated O-glycan, frequently detected in colorectal cancer (CRC), but its precise role in CRC metastasis is not well addressed. Here we investigated the effects of Core 1 β3Gal-T specific molecular chaperone (Cosmc) deletion-mediated Tn antigen exposure on CRC metastasis and its underlying mechanism. We first used CRISPR/Cas9 technology to knockout Cosmc, which is required for normal O-glycosylation, and thereby obtained Tn-positive CRC cells. We then investigated the biological consequences of Tn antigen expression in CRC. The results showed that Tn-positive cells exhibited an enhanced metastatic capability both in vitro and in vivo. A further analysis indicated that Tn antigen expression induced typical activation of epithelial-mesenchymal transition (EMT). Mechanistically, we found that H-Ras, which is known to drive EMT, was markedly up-regulated in Tn-positive cells, whereas knockdown of H-Ras suppressed Tn antigen induced activation of EMT. Furthermore, we confirmed that LS174T cells (Tn-positive) transfected with wild-type Cosmc, thus expressing no Tn antigen, had down-regulation of H-Ras expression and subsequent inhibition of EMT process. In addition, analysis of 438 samples in TCGA cohort demonstrated that Cosmc expression was reversely correlated with H-Ras, underscoring the significance of Tn antigen-H-Ras signalling in CRC patients. These data demonstrated that Cosmc deletion-mediated Tn antigen exposure promotes CRC metastasis, which is possibly mediated by H-Ras-induced EMT activation.