Merck
  • Flexible electrical aptasensor using dielectrophoretic assembly of graphene oxide and its subsequent reduction for cardiac biomarker detection.
Flexible electrical aptasensor using dielectrophoretic assembly of graphene oxide and its subsequent reduction for cardiac biomarker detection.
Scientific reports (2019-04-14)
Abhinav Sharma, Jaesung Jang
ABSTRACT
Cardiac troponin T (cTnT) is considered a clinical standard for its high specificity and sensitivity when diagnosing acute myocardial infarction; however, most studies on the electrical sensors of cardiac troponin biomarkers have focused on cTnI rather than cTnT. This study presents label-free, low-cost, transparent, and flexible aptamer-based immunosensors for the electrical detection of cTnT using reduced graphene oxide (rGO) sheets. GO was first deposited by AC dielectrophoresis between two predefined source and drain electrodes on a 3-aminopropyltriethoxylsilane-modified polyethylene terephthalate substrate. The GO was then reduced using hydrazine vapour without damaging the substrate, resulting in uniform, controlled, and stable deposition of rGO sheets, and demonstrating more stability than those directly deposited by dielectrophoresis. Amine-modified single-strand DNA aptamers against cTnT were immobilized onto the rGO channels. The relative resistance change of this sensor owing to the attachment of cTnT was quantified as the cTnT concentration decreased from 10 ng/mL to 1 pg/mL in phosphate buffered saline (PBS) and 10-fold diluted human serum in PBS, with the limits of detection being 1.2 pg/mL and 1.7 pg/mL, respectively, which is sufficiently sensitive for clinical applications. High-yield and rapid fabrication of the present rGO sensors will have significant influences on scaled-up fabrication of graphene-based sensors.