Merck
Disorder raises the critical temperature of a cuprate superconductor.
Proceedings of the National Academy of Sciences of the United States of America (2019-05-16)
Maxime Leroux, Vivek Mishra, Jacob P C Ruff, Helmut Claus, Matthew P Smylie, Christine Opagiste, Pierre Rodière, Asghar Kayani, G D Gu, John M Tranquada, Wai-Kwong Kwok, Zahirul Islam, Ulrich Welp
ABSTRACT
With the discovery of charge-density waves (CDWs) in most members of the cuprate high-temperature superconductors, the interplay between superconductivity and CDWs has become a key point in the debate on the origin of high-temperature superconductivity. Some experiments in cuprates point toward a CDW state competing with superconductivity, but others raise the possibility of a CDW-superconductivity intertwined order or more elusive pair-density waves (PDWs). Here, we have used proton irradiation to induce disorder in crystals of [Formula: see text] and observed a striking 50% increase of [Formula: see text], accompanied by a suppression of the CDWs. This is in sharp contrast with the behavior expected of a d-wave superconductor, for which both magnetic and nonmagnetic defects should suppress [Formula: see text] Our results thus make an unambiguous case for the strong detrimental effect of the CDW on bulk superconductivity in [Formula: see text] Using tunnel diode oscillator (TDO) measurements, we find indications for potential dynamic layer decoupling in a PDW phase. Our results establish irradiation-induced disorder as a particularly relevant tuning parameter for the many families of superconductors with coexisting density waves, which we demonstrate on superconductors such as the dichalcogenides and [Formula: see text].